Cerium(III) carbonate hydrate ≥99.999% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
44496-14
44496-22
44496-36
CAAAA44496-14EA
184.86
CAD
CAAAA44496-14
CAAAA44496-22
CAAAA44496-36
Cerium(III) carbonate hydrate ≥99.999% (REO, rare earth oxide basis), REacton®
Cerium (III) carbonate hydrate
Crystalline
Purity: 99.999%
CAS No.: 54451-25-1
Molecular Formula: Ce2(CO3)3.xH2O
Formula Weight: 460.27(anhy)
Physical Form: Crystalline
Melting Point: 848°
MDL No.: MFCD00149635
Formula:
C₃Ce₂O₉·H₂O Melting Pt: 848 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149635 CAS Number: 54451-25-1 EINECS: 208-655-6 Merck Index: 14,1996 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...