(S)-(+)-Methyl-3-hydroxy-2-methylpropionate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44483-04
44483-09
CAAAA44483-04EA
200.66
CAD
CAAAA44483-04
CAAAA44483-09
(S)-(+)-Methyl-3-hydroxy-2-methylpropionate ≥98%
(S)-(+)-Methyl-3-hydroxy-2-methylpropionate
Liquid
Purity: 99%
CAS No.: 80657-57-4
Molecular Formula: C5H10O3
Formula Weight: 118.13
Flash Point: 81°(178°F)
Physical Form: Liquid
Density: 1.07
Refractive Index: 1.4250
Appearance: Colorless
Boiling Point: 74°/10mm
Beilstein Reference: 1720830
Optical Rotation: +27° (c=4 in methanol)
MDL No.: MFCD00064520
Formula:
C₅H₁₀O₃ Boiling Pt: 74 °C (10 mmHg) Density: 1,07 Flash Pt: 81 °C (178 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064520 CAS Number: 80657-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...