2-(Hydroxymethyl)-18-crown-6 97%

Supplier: Thermo Scientific Chemicals
Warning

44477-01 44477-03
CAAAA44477-01EA 145.52 CAD
CAAAA44477-01 CAAAA44477-03
2-(Hydroxymethyl)-18-crown-6 97%
2-(Hydroxymethyl)-18-crown-6

Liquid


Purity: 97% CAS No.: 70069-04-4 Molecular Formula: C13H27O7 Formula Weight: 294.35 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.174 Refractive Index: 1.479 Boiling Point: 150°/0.5mm Beilstein Reference: 4313301 MDL No.: MFCD00188046

Formula: C₁₃H₂₆O₇
Boiling Pt: 150 °C (0.5 mmHg)
Density: 1,174
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00188046
CAS Number: 70069-04-4
EINECS: 274-299-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR