Cerium(IV) isopropoxide (37 - 45% Ce)
Supplier: Thermo Scientific Chemicals
|
Danger
|
44468-03
44468-06
CAAAA44468-03EA
210.14
CAD
CAAAA44468-03
CAAAA44468-06
Cerium(IV) isopropoxide (37 - 45% Ce)
Cerium(IV) isopropoxide
Powder
CAS No.: 63007-83-0
Molecular Formula: C12H28CeO4
Formula Weight: 376.48
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: Orange-red to yellow
Melting Point: 140° dec.
Solubility: Soluble in pyridine
UN No.: 2925
MDL No.: MFCD00061483
Formula:
C₁₂H₂₈CeO₄ Melting Pt: 140 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00061483 CAS Number: 63007-83-0 UN: 2925 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...