Benzyl mercaptan, polymer supported 2,0-4,0 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
44466-04
44466-09
44466-18
CAAAA44466-04EA
80.42
CAD
CAAAA44466-04
CAAAA44466-09
CAAAA44466-18
Benzyl mercaptan, polymer supported 2,0-4,0 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Benzyl mercaptan, polymer supported
Beads
Physical Form: Beads
Appearance: White to pink
MDL No.: MFCD01863867
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...