O,S-Diethyl methylphosphonothionate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
44465-02
44465-04
CAAAA44465-02EA
802.64
CAD
CAAAA44465-02
CAAAA44465-04
O,S-Diethyl methylphosphonothionate 97%
O,S-Diethyl methylphosphonothionate
Liquid
Purity: 97%
CAS No.: 2511-10-6
Molecular Formula: C5H13O2PS
Formula Weight: 168.19
Physical Form: Liquid
Density: 1.094
Refractive Index: 1.4740
Boiling Point: 95-97°/10mm
UN No.: 3278
MDL No.: MFCD01705977
Formula:
C₅H₁₃O₂PS Boiling Pt: 95…97 °C (10 mmHg) Density: 1.094 Storage Temperature: Ambient |
MDL Number:
MFCD01705977 CAS Number: 2511-10-6 UN: 3278 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...