4-Methylbenzhydrylamine hydrochloride, polymer supported 0.5 - 1.0 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
44455-06
44455-14
CAAAA44455-06EA
128.3
CAD
CAAAA44455-06
CAAAA44455-14
4-Methylbenzhydrylamine hydrochloride, polymer supported 0.5 - 1.0 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
4-Methylbenzhydrylamine hydrochloride, polymer supported
Beads
Physical Form: Beads
Appearance: Off-white to pale yellow
MDL No.: MFCD00801150
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...