Tris[N,N-bis(trimethylsilyl)amide]erbium(III) 98%

Supplier: Thermo Scientific Chemicals
Danger

44421-03 44421-06
CAAAA44421-03EA 222.78 CAD
CAAAA44421-03 CAAAA44421-06
Tris[N,N-bis(trimethylsilyl)amide]erbium(III) 98%
Tris[N,N-bis(trimethylsilyl)amide]erbium (III)

Powder- Packaged under argon


Purity: 98% CAS No.: 103457-72-3 Molecular Formula: C18H54ErN3Si6 Formula Weight: 648.41 Physical Form: Powder- Packaged under argon Sensitivity: Moisture Sensitive Appearance: Pink Melting Point: 166° UN No.: 3396 MDL No.: MFCD00271002

Formula: C₁₈H₅₄ErN₃Si₆
MW: 648.41 g/mol
Melting Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00271002
CAS Number: 103457-72-3
UN: 3396
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR