2-Chlorotrityl chloride 1.0 - 1.4 mmol/g on polystyrene, cross-linked with 1% DVB 100-200 mesh

Supplier: Thermo Scientific Chemicals

Synonyms: (2-Chlorophenyl)diphenylmethyl chloride, Chloro(2-chlorophenyl)diphenylmethane, (Chloro(2-chlorophenyl)methylene)dibenzene

44407-04 44407-09 44407-18
CAAAA44407-04EA 145.52 CAD
CAAAA44407-04 CAAAA44407-09 CAAAA44407-18
2-Chlorotrityl chloride 1.0 - 1.4 mmol/g on polystyrene, cross-linked with 1% DVB 100-200 mesh
2-Chlorotrityl chloride

Powder


CAS No.: 42074-68-0 Physical Form: Powder MDL No.: MFCD00040399

Formula: C₁₉H₁₄Cl₂
MW: 313.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040399
CAS Number: 42074-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR