Trityl chloride resin 0.5 - 2.5 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trityl chloride, polymer-supported, Chlorodiphenylmethyl Polystyrene resin
44397-03
44397-06
44397-14
CAAAA44397-03EA
67.47
CAD
CAAAA44397-03
CAAAA44397-06
CAAAA44397-14
Trityl chloride resin 0.5 - 2.5 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Trityl chloride resin
Beads
Physical Form: Beads
Sensitivity: Hygroscopic
Appearance: White to off-white
UN No.: 3261
MDL No.: MFCD00284810
Storage Temperature:
Ambient |
MDL Number:
MFCD00284810 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...