Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III) 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
44381-03
44381-06
CAAAA44381-03EA
206.98
CAD
CAAAA44381-03
CAAAA44381-06
Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III) 97%
Tris[N,N-bis(trimethylsilyl)amide]lanthanum (III)
Powder
Purity: 97%
CAS No.: 175923-07-6
Molecular Formula: C18H54LaN3Si6
Formula Weight: 620.06
Flash Point: 2°(36°F)
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 149-152°
UN No.: 3396
MDL No.: MFCD00271009
Formula:
C₁₈H₅₄LaN₃Si₆ Melting Pt: 149…152 °C Flash Pt: 2 °C (36 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00271009 CAS Number: 175923-07-6 UN: 3396 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...