5-Vinyluracil ≥97%

Supplier: Thermo Scientific Chemicals
Warning

44379-01 44379-03 44379-06
CAAAA44379-01EA 164.32 CAD
CAAAA44379-01 CAAAA44379-03 CAAAA44379-06
5-Vinyluracil ≥97%
5-Vinyluracil

Powder


Purity: 97% CAS No.: 37107-81-6 Molecular Formula: C6H6N2O2 Formula Weight: 138.12 Physical Form: Powder MDL No.: MFCD00913267

Formula: C₆H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00913267
CAS Number: 37107-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR