Bis(cyclopentadienyl)zirconium dihydride 98%

Supplier: Thermo Scientific Chemicals
Danger

44377-03 44377-06
CAAAA44377-03EA 106.97 CAD
CAAAA44377-03 CAAAA44377-06
Bis(cyclopentadienyl)zirconium dihydride 98%
Bis(cyclopentadienyl)zirconium dihydride

Powder


Hydrogenation catalyst for alkenes and alkynes


Hydrogenation catalyst for alkenes and alkynes

Formula: C₁₀H₁₂Zr
MW: 223.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04126436
CAS Number: 37342-98-6
UN: 3131
ADR: 4.3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR