Tris[N,N-bis(trimethylsilyl)amide]gadolinium(III) 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
44359-03
44359-06
CAAAA44359-03EA
154.52
CAD
CAAAA44359-03
CAAAA44359-06
Tris[N,N-bis(trimethylsilyl)amide]gadolinium(III) 98%
Tris[N,N-bis(trimethylsilyl)amide]gadolinium (III)
Powder
Purity: 98%
CAS No.: 35789-03-8
Molecular Formula: C18H54GdN3Si6
Formula Weight: 638.41
Physical Form: Powder
Sensitivity: Moisture Sensitive
UN No.: 3396
MDL No.: MFCD03427091
Formula:
C₁₈H₅₄GdN₃Si₆ Melting Pt: 161…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427091 CAS Number: 35789-03-8 UN: 3396 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...