Tris(cyclopentadienyl)holmium(III) ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Ampuled under argon
44302-03
44302-06
CAAAA44302-03EA
170.64
CAD
CAAAA44302-03
CAAAA44302-06
Tris(cyclopentadienyl)holmium(III) ≥98%
Tris(cyclopentadienyl)holmium(III)
Crystalline, Ampouled under argon
Purity: 98%
CAS No.: 1272-22-6
Molecular Formula: C15H15Ho
Formula Weight: 360.21
Physical Form: Crystalline, Ampouled under argon
Sensitivity: Moisture Sensitive
Melting Point: 295°
Boiling Point: 230° subl. (vac.)
UN No.: 3181
MDL No.: MFCD03427140
Formula:
C₁₅H₁₅Ho MW: 360.21 g/mol Boiling Pt: 230 °C (subl. in vacuum) Melting Pt: 295 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427140 CAS Number: 1272-22-6 EINECS: 215-047-4 UN: 3181 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...