Multi-element plasma standard solution, Non Metals, Specpure®, Thermo Scientific
Supplier: Thermo Scientific Chemicals
44267-AE
44267-AP
CAAAA44267-AEEA
195.92
CAD
CAAAA44267-AE
CAAAA44267-AP
Multi-element plasma standard solution, Non Metals, Specpure®, Thermo Scientific
Standards
Cationic Standards
Multi-Element Standards
As, B, P, S, Se, Si, Te at concentration 100µg/ml in 5% HCL with a trace of HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00144997
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...