Fullerene nanotube multi-walled 20-50 nm OD, Length: 5-20 micron

Supplier: Thermo Scientific Chemicals
Warning

44192-03 44192-06
CAAAA44192-03EA 339.7 CAD
CAAAA44192-03 CAAAA44192-06
Fullerene nanotube multi-walled 20-50 nm OD, Length: 5-20 micron
Fullerene nanotube

Powder


CAS No.: 308068-56-6 Physical Form: Powder Appearance: Black UN No.: 1325 MDL No.: MFCD00133992

Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 308068-56-6
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR