Amberlyst® 15 (H), wet ion exchange resin
Supplier: Thermo Scientific Chemicals
44079-30
44079-A1
44079-A7
CAAAA44079-30EA
69.05
CAD
CAAAA44079-30
CAAAA44079-A1
CAAAA44079-A7
Amberlyst® 15 (H), wet ion exchange resin
Amberlyst® 15 (H)
Beads
CAS No.: 39389-20-3
Molecular Formula: Styrene-DVB; Strongly acidic macroreticular resin
Physical Form: Beads
Sensitivity: Hygroscopic
MDL No.: MFCD00145841
Formula:
(C₁₀H₁₀·C₈H₈O₃S)x Storage Temperature: Ambient |
MDL Number:
MFCD00145841 CAS Number: 39389-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...