Ammonium magnesium phosphate hydrate ≥99.999%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44053-18
44053-30
CAAAA44053-18EA
194.34
CAD
CAAAA44053-18
CAAAA44053-30
Ammonium magnesium phosphate hydrate ≥99.999%
Ammonium magnesium phosphate hydrate
Powder
Purity: 99.999%
CAS No.: 7785-21-9
Molecular Formula: NH4MgPO4.xH2O
Formula Weight: 137.32(anhy)
Physical Form: Powder
Density: 1.711
MDL No.: MFCD00050358
Formula:
NH₄MgPO₄·xH₂O MW: 137.32 g/mol (anhy) Density: 1.711 Storage Temperature: Ambient |
MDL Number:
MFCD00050358 CAS Number: 7785-21-9 EINECS: 232-075-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...