Disposable Infrared Cards, Thermo Scientific
Supplier: Thermo Scientific Chemicals
43929-KM 43932-KJ 43932-KM 43935-KM 43936-KJ 43936-KM 43937-KJ 43937-KM 44527-KM
CAAAA43929-KJEA 283.36 CAD
CAAAA43929-KJ CAAAA43929-KM CAAAA43930-KJ CAAAA43930-KM CAAAA43931-KJ CAAAA43931-KM CAAAA43932-KJ CAAAA43932-KM CAAAA43934-KJ CAAAA43934-KM CAAAA43935-KJ CAAAA43935-KM CAAAA43936-KJ CAAAA43936-KM CAAAA43937-KJ CAAAA43937-KM CAAAA44527-KM
Disposable Infrared Cards, Thermo Scientific
Spectrophotometers and Photometers IR and FTIR Spectrophotometers
These disposable cards are specifically designed to provide a conveniently mounted non-absorbing substrate for qualitative infrared analysis.
Order Now
SPECIFICATIONS
Environmentally Preferable