(Aminoethylaminomethyl)phenethyltrimethoxysilanes mixture of m- and p-isomers

Supplier: Thermo Scientific Chemicals
Danger

43904-04 43904-09
CAAAA43904-04EA 53.4 CAD
CAAAA43904-04 CAAAA43904-09
(Aminoethylaminomethyl)phenethyltrimethoxysilanes mixture of m- and p-isomers
(Aminoethylaminomethyl)phenethyltrimethoxysilanes

Liquid


Applications: As a crosslinking agent


Applications: As a crosslinking agent

Formula: H₂NC₂H₄NHCH₂C₆H₄C₂H₄Si(OCH₃)₃
Boiling Pt: 128 °C (0.2 mmHg)
Density: 1.02
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00054775
CAS Number: 74113-77-2
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR