Aluminum oxide-Silicon oxide 13%, low surface area macroporous catalyst support
Supplier: Thermo Scientific Chemicals
43863-A1
43863-A7
CAAAA43863-A1EA
316
CAD
CAAAA43863-A1
CAAAA43863-A7
Aluminum oxide-Silicon oxide 13%, low surface area macroporous catalyst support
Aluminum oxide-Silicon oxide
3/16" Spheres
Physical Form: 3/16" Spheres
MDL No.: MFCD00058866
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...