3,3'-Diethylthiadicarbocyanine iodide [DiSC₂(5)] ≥99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DTDCI, DiSC2(5), 3-Ethyl-2-[5-(3-ethyl-2(3H)-benzothiazolylidene)-1,3-pentadienyl]benzothiazolium iodide

43851-02 43851-04 43851-09
CAAAA43851-02EA 65.1 CAD
CAAAA43851-02 CAAAA43851-04 CAAAA43851-09
3,3'-Diethylthiadicarbocyanine iodide [DiSC₂(5)] ≥99%
3,3'-Diethylthiadicarbocyanine iodide [DiSC₂(5)]

Applications: As a solvent soluble laser dye


Purity: 99% CAS No.: 514-73-8 Molecular Formula: C23H23N2S2I Formula Weight: 518.47 Physical Form: Powder Appearance: Green Melting Point: 248° dec. Solubility: Soluble in methanol Beilstein Reference: 3838938 UN No.: 2811 MDL No.: MFCD00074829


Applications: As a solvent soluble laser dye

Formula: C₂₃H₂₃N₂S₂I
MW: 518.48 g/mol
Melting Pt: 248 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00074829
CAS Number: 514-73-8
EINECS: 208-186-7
UN: 2811
ADR: 6.1,II
Merck Index: 14,3369

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR