catalyst support
Supplier: Thermo Scientific Chemicals
43815-22
43815-36
43815-A4
CAAAA43815-22EA
151.84
CAD
CAAAA43815-22
CAAAA43815-36
CAAAA43815-A4
catalyst support
Zirconium oxide
1/8" Pellets
Applications: Catalyst carrier
Applications: Catalyst carrier
Formula:
OZr MW: 107.22 Storage Temperature: Ambient |
MDL Number:
MFCD00011310 CAS Number: 12036-01-0 EINECS: 215-227-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...