Iminodiacetic acid disodium salt hydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

43809-18 43809-30
CAAAA43809-18EA 79.95 CAD
CAAAA43809-18 CAAAA43809-30
Iminodiacetic acid disodium salt hydrate 97%
Iminodiacetic acid disodium salt hydrate

Crystalline


Purity: 97% CAS No.: 17593-73-6 Molecular Formula: HN(CH2CO2Na)2.xH2O Formula Weight: 177.07(anhy) Physical Form: Crystalline

Formula: C₄H₅NNa₂O₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150689
CAS Number: 207398-95-6
EINECS: 213-181-8
Merck Index: 14,4917

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR