Cyanide, standard solution, Specpure®, CN- 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
43804-AE
43804-AP
CAAAA43804-AEEA
210.14
CAD
CAAAA43804-AE
CAAAA43804-AP
Cyanide, standard solution, Specpure®, CN- 1000 µg/ml, Thermo Scientific
Standards
Anionic Standards
Cyanide Standards
Molecular Formula: KCN in 0.1% NaOH
Physical Form: Liquid
MDL No.: MFCD00468042
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...