Ethyl-N-phenylformimidate ≥97%

Supplier: Thermo Scientific Chemicals
Warning

43788-06 43788-14
CAAAA43788-06EA 44.24 CAD
CAAAA43788-06 CAAAA43788-14
Ethyl-N-phenylformimidate ≥97%
Ethyl-N-phenylformimidate

Liquid


Purity: 97% CAS No.: 6780-49-0 Molecular Formula: C9H11NO Formula Weight: 149.19 Physical Form: Liquid Sensitivity: Air and Moisture Sensitive Appearance: Colourless liquid Boiling Point: 101-102°/15mm MDL No.: MFCD00043655

Formula: C₉H₁₁NO
Boiling Pt: 101…102 °C (15 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00043655
CAS Number: 6780-49-0
EINECS: 229-841-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR