Grinding Pieces, Zirconium oxide, Yttria stabilized, Thermo Scientific

Supplier: Thermo Scientific Chemicals
43285-30 43285-A1 43286-30 43286-A1
CAAAA43285-30EA 267.02 CAD
CAAAA43285-30 CAAAA43285-A1 CAAAA43286-30 CAAAA43286-A1
Grinding Pieces, Zirconium oxide, Yttria stabilized, Thermo Scientific
Beads Ceramic Beads
The density of the zirconium oxide beads is 6.02 g/cm³

  • MDL No.: MFCD00145546
  • Yttrium oxide (mole) 3%
  • Hardness 12
  • Total impuroty level < 0.1

Storage at ambient temperatures.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR