Potassium hydrogen diiodate 99.95-100.05% ACS primary standard

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium bi-iodate

41986-14 41986-22
CAAA41986-14EA 112.81 CAD
CAAA41986-14 CAAA41986-22
Potassium hydrogen diiodate 99.95-100.05% ACS primary standard
Potassium hydrogen diiodate

Powder


MDL: MFCD00011400

Formula: HI₂KO₆
MW: 389.91
Storage Temperature: Ambient
MDL Number: MFCD00011400
CAS Number: 13455-24-8
EINECS: 236-650-9
UN: 3085
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR