Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III) ≥99.9%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III), Bi(TMHD)3
41202-03
41202-06
CAAAA41202-03EA
117.24
CAD
CAAAA41202-03
CAAAA41202-06
CAAAA41202-MD
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III) ≥99.9%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III)
Solid
Purity: 99.9%
CAS No.: 142617-53-6
Molecular Formula: C33H57BiO6
Formula Weight: 758.79
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Appearance: White to brown
Melting Point: 114-116°
Boiling Point: 150°/0.05mm
MDL No.: MFCD00064763
Formula:
C₃₃H₅₇BiO₆ MW: 758.782 g/mol Boiling Pt: 150 °C (0.05 mmHg) Melting Pt: 114…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064763 CAS Number: 142617-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...