SP Bel-Art Poxygrid® 60- and 150-Place Test Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
Scienceware®
F18760-1150 F18751-0160 F18756-0160 F18763-0160 F18752-1150
89187-810CS 2274.31 CAD
89187-810 89187-812 89187-802 89187-804 89187-806
SP Bel-Art Poxygrid® 60- and 150-Place Test Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Chemical- and corrosion-resistant steel wire test tube racks are coated with heat-cured epoxy for long life and easy cleaning.
  • Epoxy-coated steel wire construction
  • Racks feature slotted grid bottom and reinforced edges
  • Autoclavable
  • Chemical- and corrosion-resistant
Steel wire construction with epoxy coating is unaffected by organic solvents, salts, weak acids, and weak alkalis
Racks are recommended for use within a temperature range of -210 to 121°C (-346 to 250°F).
All racks have square holes, three decks, and a slotted grid bottom and reinforced edges. Autoclavable.