KIMTECH PURE® A5 Sterile Boot Covers with CLEAN-DON™ Technology and Grasp Ties, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Microflex
31696 31697 31683
CA89203-944CS 2078.2 CAD
CA89203-944 CA89203-946 CA89203-942
KIMTECH PURE® A5 Sterile Boot Covers with CLEAN-DON™ Technology and Grasp Ties, KIMBERLY-CLARK PROFESSIONAL®
Disposable Garments
CLEAN-DON™ technology makes donning easier, combining two steps in one to save time and minimize the risk of contamination
  • Two steps in one saves time, minimizes risk of contamination
  • Edged vinyl sole to reduce slipping and liquid penetration
  • Side grasp ties for secure fit
  • Type 6 PPE Category III
  • SAL 10-6
More comfort, more efficient donning, more confidence and peace of mind. Edge vinyl sole to reduce slipping and liquid penetration. Side Grasp ties for secure fit. Type 6 PPE Category III for "parts of the body". Sterility assurance level 10-6. Vacuum-sealed packaging. White.