VWR® Standard Microplate Vortex Mixers
Supplier: VWR International
89399-880 89399-882
89399-880EA 1045.63 CAD
89399-880 89399-882
VWR® Standard Microplate Vortex Mixers
Shakers Vortex Mixers
The VWR® Standard Microplate Vortex Mixer is a variable speed analog mixer that is designed for continuous duty.
  • Designed for continuous duty
  • Designed for shaking microplates or tubes
  • Now with Best-in-Class, Exclusive 5-Year Warranty
The high speed and small orbit is optimal for effectively mixing microplates. Choose from two modes of operation: "Touch" mode which activates mixing when cup head is depressed or "On" mode when using any of the accessory attachments for continuous operation. Analog controls. Continous duty rating.
Certifications: RoHSMore Product Information