Regulator, Line, Diaphragm, Brass, Chrome Plated, Rotarex
Supplier: Rotarex
SL1544
89130-368EA 1921.91 CAD
89130-368
Regulator, Line, Diaphragm, Brass, Chrome Plated, Rotarex
Gas Regulators
Used as a line regulator for high-flow industrial or lab applications.
As a second level of regulation the S 15 will supply a precise outlet pressure to the process. Can be used for wall or panel mounting with its compact design, the rear threads and fixing ring.
Pressure control with Balanced-Valve Technology: the effect of inlet pressure fluctuations on outlet pressure are minimized. BV-technology delivers an exceptionally stable outlet pressure and flow even with high flow line regulators. Longer useful regulator lifetime and lower total ownership cost.
Ordering information: Additional outlet pressures and flow rates available; contact your Avantor sales representative for more information.