Regulator, Line, Diaphragm, Stainless Steel, Rotarex
Supplier: Rotarex
SI55145
89130-378EA 2243.55 CAD
89130-378
Regulator, Line, Diaphragm, Stainless Steel, Rotarex
Gas Regulators
Designed for line regulator applications in petrochemical, industrial and laboratory environments
With its compact design, the rear threads and its fixing ring it can be used for wall or panel mounting. Multiple mounting possibilities due to its inlet/outlet variations.
Used in calibration gas mixtures for petrochemical industry; environmental emission monitoring, industrial hygiene or safety monitors and trace impurity analyzers. Also commonly used to oxygenate fish-breeding tanks.
Ordering information: Additional outlet pressures and flow rates available; contact your Avantor sales representative for more information.