MiniVac Waste Aspiration System, Argos Technologies
Supplier: Antyila Scientific
04396-19 04396-15 04396-16 04396-17 04396-18
89401-184EA 486.47 CAD
89401-184 89401-186 89401-188 89401-190 89401-192
MiniVac Waste Aspiration System, Argos Technologies
Pumps Vacuum Aspiration Systems
MiniVac waste collection system was designed with safety, convenienc,e and bench-top space in mind
The unit consists of a disposable 1L polystyrene flask, plastic reservoir stand, polypropylene cap with barbed fittings and tubing. One tubing set is connected to primary or secondary source and one is used to aspirate the liquid. MiniVac flask and cap sit in a clear PETG plastic reservoir stand that collects liquid should flask crack or leak. Full flasks can be transferred safely by using supplied transfer cap. Small footprint (7.5"); easy clean up with 70% isopropyl alcohol. Can be usedMore Product Information