Pressure Equalizing Funnels, Ace Glass Incorporated
Supplier: Ace Glass
9490-10 7292-32 7292-30 9491-06 7291-12 9490-02 9491-03 9491-07 9490-06 7291-10 9491-05 7292-36 7291-08 7291-14 9491-15 7292-34 9490-04
89058-716EA 819.41 CAD
89058-716 89074-960 89058-704 89058-718 89074-954 89074-964 89074-952 89074-962 89074-968 89074-956 89074-966 89074-948 89058-660 89058-664 89058-720 89058-654 89058-656
Pressure Equalizing Funnels, Ace Glass Incorporated
Funnels Separating Funnels
Cylindrical pressure equalizing funnel with an outer standard taper joint top, and inner standard taper joint bottom with drip stem
The drip stem does not extend beyond joint, thereby eliminating possible source of breakage. Stopcock is available with glass plug or 1:5 PTFE plug. Fabricated from borosilicate glass.
Ordering information: Stoppers are not supplied.