Dispensing Bins, PVC/Acrylic, TrippNT
Supplier: TrippNT
50408 50160 50005
11376-204EA 269.34 CAD
11376-204 11376-206 11376-208
Dispensing Bins, PVC/Acrylic, TrippNT
Boxes Dispenser Boxes
White PVC/acrylic dispensing bins are great for the bench or countertop
Made from chemically resistant PVC and clear acrylic. The removable top allows for easy filling and the hinged clear acrylic front protects and dispenses supplies. Great for pipette tips, swabs, vials, tubes, and other small consumables.