Distillation Column, Snyder, Ace Glass Incorporated
Supplier: Ace Glass
6575-02 6575-04 6575-06
89055-650EA 361.9 CAD
89055-650 89055-652 89055-654
Distillation Column, Snyder, Ace Glass Incorporated
Distillation Components Distillation Columns
This floating ball type Snyder distillation column has a reinforced [ST] 24/40 outer joint at top and [ST] 24/40 inner joint at bottom
Each section is approximately 50 mm long. Fabricated from borosilicate glass. 89055-650 has 3 sections, 89055-652 has 6 sections, and 89055-654 has 12 sections.