Distillation Column, Vacuum Jacketed and Silvered, Ace Glass Incorporated
Supplier: Ace Glass
9335-02 9335-08
89074-406EA 1543.52 CAD
89074-406 89074-408
Distillation Column, Vacuum Jacketed and Silvered, Ace Glass Incorporated
Distillation Components Distillation Columns
This vacuum jacketed distillation column is silvered with an observation stripe
It has an enlarged bulb knockdown section that removes high boiling ends without choking. The I.D. of the column is approximately 10mm. It has indents to hold the packing. Fabricated from borosilicate glass.
Ordering information: Column is available in two lengths. An un-silvered version is available via special order.
Order Now
SPECIFICATIONS
Environmentally Preferable