Hose Connections, Regular Size, Ace Glass Incorporated
Supplier: Ace Glass
8470-03 8470-07 8470-11
89063-110PK 106.41 CAD
89063-110 89063-112 89063-114
Hose Connections, Regular Size, Ace Glass Incorporated
Fittings and Connectors
These ring-style hose connections are for use with regular size condensers
Available on straight tubing, or with a flare opposite the rings. Fabricated from borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable