p-Xylenol blue ≥95.0%

Supplier: TCI America

Synonyms: Xylenol Blue

X0016-1G X0016-25G
TCX0016-001GEA 58.46 CAD
TCX0016-001G TCX0016-025G
p-Xylenol blue ≥95.0%
p-Xylenol Blue

CAS Number: 125-31-5
MDL Number: MFCD00005870
Molecular Formula: C23H22O5S
Molecular Weight: 410.48
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: Deep Red
Lambda max.: 435 nm (pH7.0)

Formula: C₂₃H₂₂O₅S
MW: 410.49 g/mol
MDL Number: MFCD00005870
CAS Number: 125-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f7d4a34b92c24f36c0b82fda474245fee81a6dd&vid=5f7d4a34b92c24f36c0b82fda474245fee81a6dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR