Xanthotoxin ≥98.0%
Supplier: TCI America
X0009-100MG
X0009-1G
TCX0009-100MGEA
25.72
CAD
TCX0009-100MG
TCX0009-1G
Xanthotoxin ≥98.0%
Xanthotoxin
CAS Number: 298-81-7
MDL Number: MFCD00005009
Molecular Formula: C12H8O4
Molecular Weight: 216.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 147
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Other Pharmaceutical Ingredients, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: Methoxsalen, 8-Methoxypsoralen
Formula:
C₁₂H₈O₄ Melting Pt: 147 °C |
MDL Number:
MFCD00005009 CAS Number: 298-81-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...