D-Valine methyl ester hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: H-D-Val-OMe.HCl, H-D-Val-OMe·HCl

V0094-1G V0094-5G
TCV0094-1GEA 58.46 CAD
TCV0094-1G TCV0094-5G
D-Valine methyl ester hydrochloride ≥98.0%
D-Valine methyl ester hydrochloride

CAS Number: 7146-15-8
MDL Number: MFCD00237309
Molecular Formula: C6H13NO2
Molecular Weight: 167.63
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 165
Specific rotation [a]20/D: -15 deg (C=2, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Val-OMe.HCl

Formula: C₆H₁₃NO₂
Melting Pt: 165 °C
MDL Number: MFCD00237309
CAS Number: 7146-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=618146fd5692987ca7db2beb1de6b07cde4d4d95&vid=618146fd5692987ca7db2beb1de6b07cde4d4d95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR