(R)-(-)-2-Amino-3-methyl-1-butanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

V0077-5G V0077-1G
TCV0077-5GEA 158.63 CAD
TCV0077-5G TCV0077-1G
(R)-(-)-2-Amino-3-methyl-1-butanol ≥98.0% (by GC, titration analysis)
(R)-(-)-2-Amino-3-methyl-1-butanol

CAS Number: 4276-09-9
MDL Number: MFCD00064297
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Flash Point (°C): 90
Freezing point (°C): 32
Specific rotation [a]20/D: -18 deg (C=10, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-2-Amino-3-methyl-1-butanol

Formula: C₅H₁₃NO
Flash Pt: 90 °C
MDL Number: MFCD00064297
CAS Number: 4276-09-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61671e983dba8d9d58ca0cd0d7beec895073fec9&vid=61671e983dba8d9d58ca0cd0d7beec895073fec9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR