N-Vinyl-ε-caprolactam 98,0 Stabilised
Supplier: TCI America
Synonyms:
1-Vinylazepan-2-one
V0073-100G
V0073-500G
TCV0073-100GEA
57.84
CAD
TCV0073-100G
TCV0073-500G
N-Vinyl-ε-caprolactam 98,0 Stabilised
N-Vinyl-epsilon-caprolactam
(stabilized with HO-TEMPO)
CAS Number: 2235-00-9
MDL Number: MFCD00080693
Molecular Formula: C8H13NO
Molecular Weight: 139.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 36
Formula:
C₈H₁₃NO MW: 139.20 g/mol Melting Pt: 36 °C |
MDL Number:
MFCD00080693 CAS Number: 2235-00-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...