L-Valine methyl ester hydrochloride ≥98.5%
Supplier: TCI America
Synonyms:
H-Val-OMe.HCl, L-2-Aminoisovaleric Acid Methyl Ester Hydrochloride, L-Methyl valinate hydrochloride, H-Val-OMe·HCl
CAS Number: 6306-52-1
MDL Number: MFCD00012497
Molecular Formula: C6H13NO2
Molecular Weight: 167.63
Purity/Analysis Method: >98.5% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: 15.5 deg (C=2, H2O)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: L-2-Aminoisovaleric Acid Methyl Ester Hydrochloride, H-Val-OMe.HCl
Formula:
C₆H₁₄ClNO₂ MW: 167.63 g/mol |
MDL Number:
MFCD00012497 CAS Number: 6306-52-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...