3,4-Dimethoxyphenylacetone ≥97.0%

Supplier: TCI America

Synonyms: 1-(3,4-Dimethoxyphenyl)propan-2-one

V0052-25ML V0052-500ML
TCV0052-025MLEA 85.32 CAD
TCV0052-025ML TCV0052-500ML
3,4-Dimethoxyphenylacetone ≥97.0%
3,4-Dimethoxyphenylacetone

CAS Number: 776-99-8
MDL Number: MFCD00008772
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 167
Flash Point (°C): 113
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 1-(3,4-Dimethoxyphenyl)-2-propanone

Formula: C₁₁H₁₄O₃
Boiling Pt: 167 °C
Density: 1.12 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008772
CAS Number: 776-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20d6a76c6c2424dbc1fe50d317b333c0e3ae2795&vid=20d6a76c6c2424dbc1fe50d317b333c0e3ae2795&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR