Vinylene carbonate ≥97.0% stabilised
Supplier: TCI America
Synonyms:
1,3-Dioxol-2-one
(stabilized with BHT)
CAS Number: 872-36-6
MDL Number: MFCD00005380
Molecular Formula: C3H2O3
Molecular Weight: 86.05
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 60
Flash Point (°C): 73
Freezing point (°C): 21
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction), Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 1,3-Dioxol-2-one
Formula:
C₃H₂O₃ MW: 86.05 g/mol Boiling Pt: 60 °C Flash Pt: 73 °C |
MDL Number:
MFCD00005380 CAS Number: 872-36-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...