Urea ≥99.0% (by total nitrogen basis) for biochemical research

Supplier: TCI America

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

U0077-25G U0077-5G
TCU0077-25GEA 52.93 CAD
TCU0077-25G TCU0077-5G
Urea ≥99.0% (by total nitrogen basis) for biochemical research
Urea

[for Biochemical Research]
CAS Number: 57-13-6
MDL Number: MFCD00008022
Molecular Formula: CH4N2O
Molecular Weight: 60.06
Purity/Analysis Method: >99.0% (N)
Form: Crystal
Color: White
Melting point (°C): 135

Formula: NH₂CONH₂
Melting Pt: 135 °C
MDL Number: MFCD00008022
CAS Number: 57-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35e6d8f93582e894c552eefaf8c7e655b3799ab9&vid=e9715d6ac184f13efbaae1df27849cdcbb2a6823&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR